Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554042 Supplier Invitrogen™ Supplier No. PA554042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKHNQVEAAW LEGRIRQEFD KLREFLRVEE QAILDAMAEE TRQKQLLADE KMKQLTEETE VLAHEIERLQ MEMKEDDVSF LM Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

TRIM35 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM35 may play a role as a tumor suppressor, and is implicated in the cell death mechanism. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM35
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM35
Gene Accession No. Q9UPQ4
Gene Alias 0710005M05Rik; A430106H13Rik; AW046487; Hemopoietic lineage switch protein 5; HLS5; KIAA1098; LOW QUALITY PROTEIN: tripartite motif-containing protein 35; macrophage-derived apoptosis-inducing RBCC protein; Mair; MGC17233; mKIAA1098; NC8; Protein MAIR; Protein Nc8; Trim35; tripartite motif containing 35; tripartite motif protein 35; tripartite motif-containing 35; tripartite motif-containing protein 35
Gene Symbols TRIM35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM35. Recombinant protein control fragment (Product #RP-92095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.