Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142052
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142052 Supplier Invitrogen™ Supplier No. PA5142052
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

TRIM35 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM35 may play a role as a tumor suppressor, and is implicated in the cell death mechanism. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM35
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TRIM35
Gene Accession No. Q9UPQ4
Gene Alias 0710005M05Rik; A430106H13Rik; AW046487; Hemopoietic lineage switch protein 5; HLS5; KIAA1098; LOW QUALITY PROTEIN: tripartite motif-containing protein 35; macrophage-derived apoptosis-inducing RBCC protein; Mair; MGC17233; mKIAA1098; NC8; Protein MAIR; Protein Nc8; Trim35; tripartite motif containing 35; tripartite motif protein 35; tripartite motif-containing 35; tripartite motif-containing protein 35
Gene Symbols TRIM35
Host Species Goat
Immunogen Peptide with sequence ERSPDVSPGPSRS-C (aa 2-14).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23087
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.