Learn More
Invitrogen™ TRIM36 Polyclonal Antibody

Description
Immunogen sequence: MYCELCRRPVC HLCKLGGNHA NHRVTTMSSA YKTLKEKLSK DIDYLIGKES QVKSQISELN LLMKETECNG ERAKEEAITH FEKLFEVL Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Specifications
Specifications
| Antigen | TRIM36 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRIM36 |
| Gene Accession No. | Q9NQ86 |
| Gene Alias | Acrosome RBCC protein; D18Wsu100e; E3 ubiquitin-protein ligase Trim36; haprin; RBCC728; RING finger protein 98; RING-type E3 ubiquitin transferase TRIM36; RNF98; trim36; tripartite motif containing 36; tripartite motif protein 36; tripartite motif-containing 36; tripartite motif-containing protein 36; wu:fj48f01; zgc:153447; zgc:63476; Zinc-binding protein Rbcc728 |
| Gene Symbols | TRIM36 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.