Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM37 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059820AF647
Description
TRIM37 Polyclonal antibody specifically detects TRIM37 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TRIM37 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
EC 6.3.2.-, KIAA0898Tripartite motif-containing protein 37, Mulibrey nanism protein, MULRING-B-box-coiled-coil protein, POB1E3 ubiquitin-protein ligase TRIM37, TEF3, tripartite motif containing 37, tripartite motif-containing 37 | |
The immunogen recognized by this antibody maps to a region between residue 850 and 900 of human tripartite motif-containing 37 using the numbering given in entry NP_056109.1 (GeneID 4591). | |
0.1 mL | |
Stem Cell Markers | |
4591 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction