Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM37 Monoclonal Antibody (2D11)
GREENER_CHOICE

Catalog No. PIMA519380
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519380 Supplier Invitrogen™ Supplier No. MA519380
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GHLEGLQMTD LENNSETGEL QPVLPEGASA APEEGMSSDS DIECDTENEE QEEHTSVGGF HDSFMVMTQP PDEDTHSSFP DGEQIGPEDL SFNTDENSGR.

Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM37
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TRIM37
Gene Accession No. O94972
Gene Alias 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; E3 ubiquitin-protein ligase TRIM37; Kiaa0898; mKIAA0898; MUL; mulibrey nanism protein; POB1; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TEF3; TRAF encompassing factor 3; Trim37; tripartite motif containing 37; tripartite motif protein 37; tripartite motif-containing 37; tripartite motif-containing protein 37
Gene Symbols TRIM37
Host Species Mouse
Immunogen TRIM37 (NP_056109, 865 a.a. ~ 964 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4591
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.