Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110519 Supplier Invitrogen™ Supplier No. PA5110519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGMELLAKSS VANCYIRNST NKKSNSPKPA RSSVAGSLSL RRAVDPGENS RSKGDCQTLS EGSPGSSQSG SRHSSPRALI HGSIGDILPK TEDRQCKALD SDAVVVAVFS GLPAVEKRRK MVTLGANAKG GHLEGLQMTD LENNSETGEL QPVLPEGASA APEEGMSSDS DIECDTENEE QEEHTSVGGF HDSFMVMTQP PDEDTHSSFP DGEQIGPEDL SFNTDENSGR.

Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM37
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRIM37
Gene Accession No. Q6PCX9
Gene Alias 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; E3 ubiquitin-protein ligase TRIM37; Kiaa0898; mKIAA0898; MUL; mulibrey nanism protein; POB1; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TEF3; TRAF encompassing factor 3; Trim37; tripartite motif containing 37; tripartite motif protein 37; tripartite motif-containing 37; tripartite motif-containing protein 37
Gene Symbols TRIM37
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 735-964 of human TRIM37 (NP_0010052071).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360592, 68729
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.