Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM38 Antibody (OTI113D3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274610F
Description
TRIM38 Monoclonal specifically detects TRIM38 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TRIM38 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TRIM38 | |
Recombinant protein fragment corresponding to amino acids 1-265 of human TRIM38 (NP_006346) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Monkey | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI113D3 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet | |
Mouse | |
Affinity Purified | |
Signal Transduction, Zinc Finger | |
10475.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction