Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM38 Antibody (OTI113D3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246215
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
TRIM38 Monoclonal specifically detects TRIM38 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
TRIM38 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150, Immunofluorescence 1:100 | |
MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet | |
Mouse | |
53.2 kDa | |
0.1 mL | |
Signal Transduction, Zinc Finger | |
10475 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
OTI113D3 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
TRIM38 | |
Human recombinant protein fragment corresponding to amino acids 1-265 of human TRIM38(NP_006346) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction