Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540972 Supplier Invitrogen™ Supplier No. PA540972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRPNKQLGSL IEALKETDQE MSCEEHGEQF HLFCEDEGQL ICWRCERAPQ Sequence homology: Cow: 77%; Dog: 92%; Goat: 77%; Horse: 77%; Human: 100%; Rabbit: 85%.

This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM38
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM38
Gene Accession No. O00635
Gene Alias E3 ubiquitin-protein ligase TRIM38; Gm23; hypothetical protein LOC531022; MGC8946; RING finger protein 15; RING-type E3 ubiquitin transferase TRIM38; RNF15; Ro/SSA ribonucleoprotein homolog; RORET; TRIM38; tripartite motif containing 38; tripartite motif-containing 38; tripartite motif-containing protein 38; tripartite motif-containing protein 38; E3 ubiquitin-protein ligase TRIM38; tripartite motif-containing protein 38; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM38; zinc finger protein RoRet
Gene Symbols TRIM38
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TRIM38 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10475
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.