Learn More
TRIM39 Mouse anti-Human, Clone: OTI2A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505761

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.Specifications
TRIM39 | |
Monoclonal | |
Unconjugated | |
TRIM39 | |
RNF23, TFP, TRIM39B | |
Mouse | |
Affinity Chromatography | |
RUO | |
56658 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HCM9 | |
TRIM39 | |
Human recombit protein fragment corresponding to amino acids 144-378 of human TRIM39 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.