Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582676
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582676 Supplier Invitrogen™ Supplier No. PA582676
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSQRNLVAKM KKVMHLQDVE VKNATQWKDK IKSQRMRIST EFSKLHNFLV EEEDLFLQRL NKEEEETKKK LNENTLKLNQ TIASLKKLIL EVGEKSQAPT LELLQNPK Highest antigen sequence indentity to the following orthologs: Mouse - 38%, Rat - 36%.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternative splicing of this gene generates two transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM4
Gene Accession No. Q9C037
Gene Alias E3 ubiquitin-protein ligase TRIM4; RING finger protein 87; RING-type E3 ubiquitin transferase TRIM4; RNF87; TRIM4; tripartite motif containing 4; tripartite motif protein 4; tripartite motif-containing 4; Tripartite motif-containing protein 4
Gene Symbols TRIM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM4. Recombinant protein control fragment (Product #RP-92756).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.