Learn More
Invitrogen™ TRIM40 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Specifications
Specifications
| Antigen | TRIM40 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | TRIM40 |
| Gene Accession No. | Q6P9F5 |
| Gene Alias | Probable E3 NEDD8-protein ligase; RING finger protein 35; ring finger RNF35; RNF35; TRIM40; tripartite motif containing 40; tripartite motif protein 40; tripartite motif-containing 40; tripartite motif-containing protein 40 |
| Gene Symbols | TRIM40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.