Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560525 Supplier Invitrogen™ Supplier No. PA560525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQWLGQLEHM PAEAARILDI SRAVTQLRSL VIDLERTAKE LDTNTLKNAG DLLNRSAPQK LEVIYPQLEK GVSELLLQP Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.

This gene encodes a member of the tripartite motif (TRIM) protein family. The encoded protein may play a role as a negative regulator against inflammation and carcinogenesis in the gastrointestinal tract. Alternatively spliced transcript variants that encode different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM40
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM40
Gene Accession No. Q6P9F5
Gene Alias Probable E3 NEDD8-protein ligase; RING finger protein 35; ring finger RNF35; RNF35; TRIM40; tripartite motif containing 40; tripartite motif protein 40; tripartite motif-containing 40; tripartite motif-containing protein 40
Gene Symbols TRIM40
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM40. Recombinant protein control fragment (Product #RP-99565).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135644
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.