Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540900 Supplier Invitrogen™ Supplier No. PA540900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AQSSTEQTLL SPSEKPRRFG VYLDYEAGRL GFYNAETLAH VHTFSAAFLG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ). See TRIM45.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM41
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM41
Gene Accession No. Q8WV44
Gene Alias AW552703; BC020156; E3 ubiquitin-protein ligase TRIM41; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM41; R75223; RINCK; RING finger-interacting protein with C kinase; RING-finger protein that interacts with C kinase; RING-type E3 ubiquitin transferase TRIM41; Trim41; tripartite motif containing 41; tripartite motif protein 41; tripartite motif-containing 41; tripartite motif-containing protein 41
Gene Symbols TRIM41
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TRIM41 (aa 561-610).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90933
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.