Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555013 Supplier Invitrogen™ Supplier No. PA555013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RESTHHKEKV GPGGSSVGSG DASSSRHHHR RRRLHLPQQP LLQREVWCVG TNGKRYQAQS STEQTLLSPS EKPRRFGVYL DYEA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ). See TRIM45.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM41
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM41
Gene Accession No. Q8WV44
Gene Alias AW552703; BC020156; E3 ubiquitin-protein ligase TRIM41; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM41; R75223; RINCK; RING finger-interacting protein with C kinase; RING-finger protein that interacts with C kinase; RING-type E3 ubiquitin transferase TRIM41; Trim41; tripartite motif containing 41; tripartite motif protein 41; tripartite motif-containing 41; tripartite motif-containing protein 41
Gene Symbols TRIM41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM41. Recombinant protein control fragment (Product #RP-93582).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90933
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.