Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM43B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571069
Description
This target displays homology in the following species: Human: 100%.
TRIM43B is a protein coding gene. Gene Ontology (GO) annotations related to this gene include zinc ion binding.
Specifications
TRIM43B | |
Polyclonal | |
Unconjugated | |
TRIM43B | |
TRIM43B; tripartite motif containing 43B; tripartite motif-containing 43B; tripartite motif-containing protein 43B; Tripartite motif-containing protein 43-like protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
653192 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6NCK2 | |
TRIM43B | |
synthetic peptide directed towards the N-terminal region of human TRIM43B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction