Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM44 Monoclonal Antibody (OTI5C2), TrueMAB™, OriGene
SDP

Catalog No. 501676016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6016 100 μL
50-253-9621 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-6016 Supplier OriGene Supplier No. TA505560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.

Specifications

Antigen TRIM44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TRIM44
Gene Accession No. Q96DX7
Gene Alias AN3, DIPB, HSA249128, MC7
Gene Symbols TRIM44
Host Species Mouse
Immunogen Full length human recombit protein of human TRIM44 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54765
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.