Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM44 Mouse anti-Human, Clone: 1B12H12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | TRIM44 |
|---|---|
| Clone | 1B12H12 |
| Concentration | 1.85 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6682
![]()
|
Proteintech Group Inc
662491IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.Specifications
| TRIM44 | |
| 1.85 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DIPB, HSA249128, MC7, Protein DIPB, TRIM44, tripartite motif containing 44 | |
| TRIM44 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1B12H12 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96DX7 | |
| 54765 | |
| TRIM44 Fusion Protein Ag18771 | |
| Primary | |
| -20°C | |
| TRIM44 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title