Learn More
TRIM44 Mouse anti-Human, Clone: OTI1E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505561

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.Specifications
TRIM44 | |
Monoclonal | |
Unconjugated | |
TRIM44 | |
AN3, DIPB, HSA249128, MC7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
54765 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96DX7 | |
TRIM44 | |
Full length human recombit protein of human TRIM44 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.