Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563459 Supplier Invitrogen™ Supplier No. PA563459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVERLKFKSS DPKVTRDQMK MFIQQEFKKV QKVIADEEQK ALHLVDIQEA MATAHVTEIL ADIQSHMDRL MTQMAQAKEQ LDTSNESAEP KAEGDEEGPS GA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 96%.

May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM44
Gene Accession No. Q96DX7
Gene Alias brain cDNA 7; DIPB; DIPB protein; HSA249128; Mc7; Protein DIPB; Protein Mc7; Trim44; tripartite motif containing 44; tripartite motif protein 44; tripartite motif-containing 44; tripartite motif-containing protein 44
Gene Symbols TRIM44
Host Species Rabbit
Immunogen Recombinant protein fragment of Human TRIM44 (Product #RP-97434)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.