Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM44 Polyclonal Antibody
TRIM44 Polyclonal Antibody
Supplier: Thermo Scientific PA570265
Description
TRIM44 Polyclonal Antibody for Western Blot

Specifications
TRIM44 | |
Polyclonal | |
Unconjugated | |
TRIM44 | |
brain cDNA 7; DIPB; DIPB protein; HSA249128; Mc7; Protein DIPB; Protein Mc7; Trim44; tripartite motif containing 44; tripartite motif protein 44; tripartite motif-containing 44; tripartite motif-containing protein 44 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
80985 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9QXA7 | |
TRIM44 | |
synthetic peptide directed towards the middle region of human TRIM44 (aa 297-346). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction