Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM44 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116073
Description
Antibody detects endogenous levels of total TRIM44.
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
Specifications
TRIM44 | |
Polyclonal | |
Unconjugated | |
TRIM44 | |
brain cDNA 7; DIPB; DIPB protein; HSA249128; Mc7; Protein DIPB; Protein Mc7; Trim44; tripartite motif containing 44; tripartite motif protein 44; tripartite motif-containing 44; tripartite motif-containing protein 44 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
362172, 54765, 80985 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q6QA27, Q96DX7, Q9QXA7 | |
TRIM44 | |
A synthesized peptide derived from human TRIM44(Accession Q96DX7), corresponding to amino acid residues E154-I204. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction