Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 50555448 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-555-448 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-555-448 Supplier Proteintech Group Inc Supplier No. 115111AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This is a rabbit polyclonal antibody raised against full length of human TRIM44.

May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.

Specifications

Antigen TRIM44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TRIM44
Gene Accession No. Q6QA27, Q96DX7, Q9QXA7
Gene Alias DIPB, HSA249128, MC7, Protein DIPB, TRIM44, tripartite motif containing 44
Gene Symbols Trim44
Host Species Rabbit
Immunogen TRIM44 Fusion Protein Ag2070
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362172, 54765, 80985
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.