Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM44 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572058
Description
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
Specifications
TRIM44 | |
Polyclonal | |
Unconjugated | |
TRIM44 | |
Tripartite motif-containing protein 44; Protein DIPB; TRIM44; DIPB | |
Rabbit | |
Protein A | |
RUO | |
54765, 80985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
Q96DX7, Q9QXA7 | |
TRIM44 | |
KLH conjugated synthetic peptide between 298-327 amino acids from the C-terminal region of human TRIM44 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction