Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM45 Antibody (OTI3G3), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274612JF549
Description
TRIM45 Monoclonal specifically detects TRIM45 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TRIM45 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRIM45 | |
Recombinant protein fragment corresponding to amino acids 288-580 of human TRIM45 (NP_079464) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3G3 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
RING finger protein 99, RNF99FLJ13181, tripartite motif containing 45, tripartite motif-containing 45, tripartite motif-containing protein 45 | |
Mouse | |
Affinity Purified | |
Apoptosis, Zinc Finger | |
80263.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction