Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM45 Antibody (OTI3G3), Novus Biologicals™
SDP

Catalog No. NBP246223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP246223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP246223 Supplier Novus Biologicals Supplier No. NBP246223
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Immunohistochemistry, Immunofluorescence

TRIM45 Monoclonal specifically detects TRIM45 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence.

Specifications

Antigen TRIM45
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3G3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:150, Immunofluorescence 1:100
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias RING finger protein 99, RNF99FLJ13181, tripartite motif containing 45, tripartite motif-containing 45, tripartite motif-containing protein 45
Gene Symbols TRIM45
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 288-580 of human TRIM45(NP_079464) produced in E.coli.
Molecular Weight of Antigen 64.2 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 80263
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.