Learn More
Invitrogen™ TRIM45 Monoclonal Antibody (OTI4F1)

Description
TRIM45 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TRIM45 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4F1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TRIM45 |
| Gene Accession No. | Q9H8W5 |
| Gene Alias | 4921530N01Rik; FLJ13181; RING finger protein 99; RNF99; TRIM45; tripartite motif containing 45; tripartite motif protein 45; tripartite motif-containing 45; tripartite motif-containing protein 45 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.