Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556420 Supplier Invitrogen™ Supplier No. PA556420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLFCEFCDRP VCQDCVVGEH REHPCDFTSN VIHKHGDSVW ELLKGTQPHV EALEEALAQI HIINSALQKR VEAVAADVR Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

TRIM45 is a member of the tripartite motif family. It may function as a transcriptional repressor of the mitogen-activated protein kinase pathway. Alternatively spliced transcript variants have been described. TRIM45 belongs to a family of tripartite motif (TRIM) proteins that play important roles in a variety of cellular functions, including cell proliferation, differentiation, development, oncogenesis, and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM45
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM45
Gene Accession No. Q9H8W5
Gene Alias 4921530N01Rik; FLJ13181; RING finger protein 99; RNF99; TRIM45; tripartite motif containing 45; tripartite motif protein 45; tripartite motif-containing 45; tripartite motif-containing protein 45
Gene Symbols TRIM45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM45. Recombinant protein control fragment (Product #RP-94211).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80263
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.