Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM46 Monoclonal Antibody (3G11)
GREENER_CHOICE

Catalog No. PIMA522628
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522628 Supplier Invitrogen™ Supplier No. MA522628
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RLPPHSPPAW HYTVEFRRTD VPAQPGPTRW QRREEVRGTS ALLENPDTGS VYVLRVRGCN KAGYGEYSED VHLHTPPAPV LHFFLDSRWG ASRERLAISK DQRAVRSVPG.

Microtubule-associated protein that is involved in the formation of parallel microtubule bundles linked by cross-bridges in the proximal axon. Required for the uniform orientation and maintenance of the parallel microtubule fascicles, which are important for efficient cargo delivery and trafficking in axons. Thereby also required for proper axon specification, the establishment of neuronal polarity and proper neuronal migration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM46
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3G11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TRIM46
Gene Accession No. Q7Z4K8
Gene Alias Gene Y protein; geneY; TRIFIC; TRIM46; tripartite motif containing 46; tripartite motif protein 46; tripartite motif-containing 46; tripartite motif-containing protein 46; tripartite, fibronectin type III and C-terminal SPRY motif-containing; tripartite, fibronectin type-III and C-terminal SPRY motif protein
Gene Symbols TRIM46
Host Species Mouse
Immunogen TRIM46 (NP_079334, 451 a.a. ~ 560 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80128
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.