Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556457 Supplier Invitrogen™ Supplier No. PA556457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MCPDHKEEVT HYCKTCQRLV CQLCRVRRTH SGHKITPVLS AYQALKDKLT KSLTYILGNQ DTVQTQICEL EEAVRHTEVS GQQAKEEVSQ LVRGLGAVLE EKR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Microtubule-associated protein that is involved in the formation of parallel microtubule bundles linked by cross-bridges in the proximal axon. Required for the uniform orientation and maintenance of the parallel microtubule fascicles, which are important for efficient cargo delivery and trafficking in axons. Thereby also required for proper axon specification, the establishment of neuronal polarity and proper neuronal migration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM46
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM46
Gene Accession No. Q7Z4K8
Gene Alias Gene Y protein; geneY; TRIFIC; TRIM46; tripartite motif containing 46; tripartite motif protein 46; tripartite motif-containing 46; tripartite motif-containing protein 46; tripartite, fibronectin type III and C-terminal SPRY motif-containing; tripartite, fibronectin type-III and C-terminal SPRY motif protein
Gene Symbols TRIM46
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM46. Recombinant protein control fragment (Product #RP-94431).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.