Learn More
Invitrogen™ TRIM46 Polyclonal Antibody

Description
Immunogen sequence: LLENPDTGSV YVLRVRGCNK AGYGEYSEDV HLHTPPAPVL HFFLDSRWGA SRERLAISKD QRAVRSVPGL PLLLAADRLL TGCHLSVDVV LGDVAVTQGR SYW Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.
Specifications
Specifications
| Antigen | TRIM46 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRIM46 |
| Gene Accession No. | Q7Z4K8 |
| Gene Alias | Gene Y protein; geneY; TRIFIC; TRIM46; tripartite motif containing 46; tripartite motif protein 46; tripartite motif-containing 46; tripartite motif-containing protein 46; tripartite, fibronectin type III and C-terminal SPRY motif-containing; tripartite, fibronectin type-III and C-terminal SPRY motif protein |
| Gene Symbols | TRIM46 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.