Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM46 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31016225UL
Description
TRIM46 Polyclonal specifically detects TRIM46 in Mouse samples. It is validated for Western Blot.Specifications
TRIM46 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
FLJ23229, Gene Y protein, GeneY, TRIFICGENEY, tripartite motif containing 46, tripartite motif-containing 46, tripartite motif-containing protein 46, Tripartite, fibronectin type-III and C-terminal SPRY motif protein | |
The immunogen is a synthetic peptide directed towards the middle region of mouse TRIM46 (NP_898858.1). Peptide sequence ASYLVKVGVGLESKLQESFQGAPDVISPRYDPDSGHDSGAEDAAVEALPP | |
25 μg | |
Zinc Finger | |
80128 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction