Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM47 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059841
Description
TRIM47 Polyclonal specifically detects TRIM47 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
TRIM47 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
Gene overexpressed in astrocytoma protein, GOARING finger protein 100, RNF100tripartite motif-containing protein 47, tripartite motif containing 47, tripartite motif-containing 47 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
NP_258411.2 | |
TRIM47 | |
The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human tripartite motif-containing 47 (gene over expressed in astrocytoma) using the numbering given in entry NP_258411.2 (GeneID 91107). | |
0.1 mL | |
Zinc Finger | |
91107 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction