Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553258 Supplier Invitrogen™ Supplier No. PA553258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQEPYDRGRL GRNAHSCCLQ WNGRSFSVWF HGLEAPLPHP FSPTVGVCLE YADRALAFYA VRDGKMSLLR RLKASRPRRG GIPASPIDPF QSRLDSHFAG LFTHRLK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

E3 ubiquitin-protein ligase that mediates the ubiquitination and proteasomal degradation of CYLD. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM47
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM47
Gene Accession No. Q96LD4
Gene Alias 2210023F24Rik; E3 ubiquitin-protein ligase TRIM47; Gene overexpressed in astrocytoma protein; GOA; RING finger protein 100; RNF100; Trim47; tripartite motif containing 47; tripartite motif protein 47; tripartite motif-containing 47; tripartite motif-containing protein 47
Gene Symbols TRIM47
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM47. Recombinant protein control fragment (Product #RP-90757).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.