Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110521 Supplier Invitrogen™ Supplier No. PA5110521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELSFTKSSQA VRAVRDMLAV ACVNQWEQLR GPGGNEDGPQ KLDSEADAEP QDLESTNLLE SEAPRDYFLK FAYIVDLDSD TADKFLQLFG TKGVKRVLCP INYPLSPTRF THCEQVLGEG ALDRGTYYWE VEIIEGWVSM GVMAEDFSPQ EPYDRGRLGR NAHSCCLQWN GRSFS.

E3 ubiquitin-protein ligase that mediates the ubiquitination and proteasomal degradation of CYLD. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM47
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TRIM47
Gene Accession No. Q8C0E3, Q96LD4
Gene Alias 2210023F24Rik; E3 ubiquitin-protein ligase TRIM47; Gene overexpressed in astrocytoma protein; GOA; RING finger protein 100; RNF100; Trim47; tripartite motif containing 47; tripartite motif protein 47; tripartite motif-containing 47; tripartite motif-containing protein 47
Gene Symbols TRIM47
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 356-530 of human TRIM47 (NP_258411.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217333, 690374, 91107
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.