Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM47 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987465
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7465 Supplier Bioss Supplier No. BS6744R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.

Specifications

Antigen TRIM47
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRIM47
Gene Accession No. Q96LD4
Gene Alias 2210023F24Rik; E3 ubiquitin-protein ligase TRIM47; Gene overexpressed in astrocytoma protein; GOA; RING finger protein 100; RNF100; TRIM47; tripartite motif containing 47; tripartite motif protein 47; tripartite motif-containing 47; Tripartite motif-containing protein 47
Gene Symbols TRIM47
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRIM47.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91107
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.