Learn More
Invitrogen™ TRIM49 Monoclonal Antibody (2E5)

Description
Peptide Sequence: ILHRSESVLL HMPQPLNPEL SAGPITGLRD RLNQFRVHIT LHHEEANNDI FLYEILRSMC IGCDHQDVPY FTATPRSFLA WGVQTFTSGK.
Specifications
Specifications
| Antigen | TRIM49 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 2E5 |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TRIM49 |
| Gene Accession No. | P0CI25 |
| Gene Alias | RING finger protein 18; RNF18; testis-specific RING Finger protein; testis-specific ring-finger protein; TRIM49; TRIM49A; TRIM49L2; tripartite motif containing 49; tripartite motif-containing 49; tripartite motif-containing protein 49 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.