Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM49C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542375
Description
Peptide sequence: QCSECTKSTE QINLKTNIHL KKMASLARKV SLWLFLSSEE QMCGTHRETK Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
| TRIM49C | |
| Polyclonal | |
| Unconjugated | |
| TRIM49C | |
| TRIM49C; TRIM49L2; tripartite motif containing 49C; tripartite motif containing 49-like 2; Tripartite motif-containing protein 49C; tripartite motif-containing protein 49-like protein 2; Tripartite motif-containing protein LOC642612 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 642612 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P0CI26 | |
| TRIM49C | |
| Synthetic peptide directed towards the N-terminal of human TRIM49C (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction