Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM5 alpha/gamma/delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518156
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518156 Supplier Invitrogen™ Supplier No. PA518156
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

TRIM5a is a 493 amino acid member of the large tripartite motif protein (TRIM) family. TRIM proteins are composed of three zinc-binding domains, a RING, a B-box 2 and a coiled-coil domain, and they use homomultimerization to identify different cell compartments. Some TRIM proteins, such as TRIM5a, also possess a carboxy-terminal B30.2 (SPRY) domain and localize to the cytoplasm. TRIM5a mediates innate intracellular retroviral resistance, which is dependent on its carboxy-terminal domain. The three variable regions of the B30.2 domain form loops on one side of the B30.2 core structure of TRIM5a which may form a binding surface for the virus. TRIM5a trimerization plays a major role in its affinity for the retroviral capsid, and in its ability to inhibit virus infection. The linker region between the coiled-coil and B30.2 domains of TRIM5a is required for this trimerization. TRIM5a blocks infection after the virus has entered the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM5 alpha/gamma/delta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TRIM5
Gene Accession No. Q9C035
Gene Alias EG667823; Gm8833; ring finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; Trim5; trim5a; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing 5; tripartite motif-containing protein 5
Gene Symbols TRIM5
Host Species Goat
Immunogen Synthetic peptide sequence (ASGILVNVKEEVTC) corresponding to the N-terminus amino acids of TRIM5 (aa 2-15).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85363
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.