Learn More
TRIM5 delta Polyclonal Antibody

Description
TRIM5 delta Polyclonal Antibody for Western Blot, IHC
Specifications
Specifications
| Antigen | TRIM5 delta |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TRIM5 |
| Gene Accession No. | Q9C035 |
| Gene Alias | EG667823; Gm8833; RING finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; TRIM5; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing 5; tripartite motif-containing protein 5 |
| Gene Symbols | TRIM5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.