Learn More
Invitrogen™ TRIM50 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144038
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human K562 whole cell, human 293T whole cell, human U-87MG whole cell. Flow: CACO-2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TRIM50 belongs to the TRIM/RBCC family. It contains 1 B box-type zinc finger, 1 B30.2/SPRY domain and 1 RING-type zinc finger. TRIM50 is an E3 ubiquitin-protein ligase.
Specifications
| TRIM50 | |
| Polyclonal | |
| Unconjugated | |
| TRIM50 | |
| E3 ubiquitin-protein ligase TRIM50; RING-type E3 ubiquitin transferase TRIM50; TRIM50; TRIM50A; tripartite motif containing 50; tripartite motif protein 50; tripartite motif-containing 50; tripartite motif-containing 50A; tripartite motif-containing protein 50 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 135892 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q86XT4 | |
| TRIM50 | |
| E.coli-derived human TRIM50 recombinant protein (Position: K243-L487). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.