Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554100 Supplier Invitrogen™ Supplier No. PA554100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFSPISFKPG LHQADIKLTV WKRLFRKVLP APEPLKLDPA TAHPLLELSK GNTVVQCGLL AQRRASQPER FDYSTC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

TRIM50 belongs to the TRIM/RBCC family. It contains 1 B box-type zinc finger, 1 B30.2/SPRY domain and 1 RING-type zinc finger. TRIM50 is an E3 ubiquitin-protein ligase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM50
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM50
Gene Accession No. Q86XT4
Gene Alias E3 ubiquitin-protein ligase TRIM50; RING-type E3 ubiquitin transferase TRIM50; TRIM50; TRIM50A; tripartite motif containing 50; tripartite motif protein 50; tripartite motif-containing 50; tripartite motif-containing 50A; tripartite motif-containing protein 50
Gene Symbols TRIM50
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM50. Recombinant protein control fragment (Product #RP-92747).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.