Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM50 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988858
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8858 Supplier Bioss Supplier No. BS9152R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM50 belongs to the TRIM/RBCC family. It contains 1 B box-type zinc finger, 1 B30.2/SPRY domain and 1 RING-type zinc finger. TRIM50 is an E3 ubiquitin-protein ligase.

Specifications

Antigen TRIM50
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRIM50
Gene Accession No. Q86XT4
Gene Alias E3 ubiquitin-protein ligase TRIM50; RING-type E3 ubiquitin transferase TRIM50; TRIM50; TRIM50A; tripartite motif containing 50; tripartite motif-containing 50A; Tripartite motif-containing protein 50
Gene Symbols TRIM50
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRIM50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135892
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.