Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM58 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554856 Supplier Invitrogen™ Supplier No. PA554856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAGEISFYNV TDGSYIYTFN QLFSGLLRPY FFICDATPLI LPPTTIAGSG NWASRDHLDP ASDVRDDHL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 71%.

E3 ubiquitin ligase induced during late erythropoiesis. Directly binds and ubiquitinates the intermediate chain of the microtubule motor dynein (DYNC1LI1/DYNC1LI2), stimulating the degradation of the dynein holoprotein complex. May participate in the erythroblast enucleation process through regulation of nuclear polarization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM58
Applications Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM58
Gene Accession No. Q8NG06
Gene Alias BIA2; E3 ubiquitin-protein ligase TRIM58; Gm16; Gm1783; Protein BIA2; RGD1566174; RING-type E3 ubiquitin transferase TRIM58; TRIM58; tripartite motif containing 58; tripartite motif-containing 58; tripartite motif-containing protein 58
Gene Symbols TRIM58
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM58. Recombinant protein control fragment (Product #RP-93602).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.