Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106512 Supplier Invitrogen™ Supplier No. PA5106512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRIM59.

May serve as a multifunctional regulator for innate immune signaling pathways. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM59
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRIM59
Gene Accession No. Q8IWR1, Q922Y2
Gene Alias 2310035M22Rik; 2700022F13Rik; IFT80L; MRF1; RGD1311956; RING finger 1; RING finger protein 1; RING finger protein 104; RNF104; TRIM57; TRIM59; tripartite motif containing 59; tripartite motif-containing 57; tripartite motif-containing 59; tripartite motif-containing protein 59; tripartite motif-containing protein 59-like protein; TSBF1; tumor suppressor TSBF1; Tumor suppressor TSBF-1; tumor suppressor TSBF1 homolog; Unknown (protein for MGC:137058)
Gene Symbols TRIM59
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRIM59(Accession Q8IWR1), corresponding to amino acid residues K201-L251.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286827, 365813, 66949
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.