Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563865 Supplier Invitrogen™ Supplier No. PA563865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQSYWVDVTL NPHTANLNLV LAKNRRQVRF VGAKVSGPSC LEKHYDCSVL GSQHFS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

TRIM6 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the nucleus, but its specific function has not been identified.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the nucleus, but its specific function has not been identified. This gene is mapped to chromosome 11p15, where it resides within a TRIM gene cluster. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A read-through transcript transcribed from this gene and TRIM34 has been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trim6
Gene Accession No. Q9C030
Gene Alias C430046K18Rik; D7Ertd684e; RING finger protein 89; RING-type E3 ubiquitin transferase TRIM6; RNF89; TRIM6; tripartite motif containing 6; tripartite motif protein 6; tripartite motif-containing 6; tripartite motif-containing protein 6
Gene Symbols Trim6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM6. Recombinant protein control fragment (Product #RP-100872).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.