Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM60 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557407 Supplier Invitrogen™ Supplier No. PA557407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYHREILEGS LEPLRNNIER VEKVIILQGS KSVELKKKVE YKREEINSEF EQIRLFLQNE QEMILRQIQD EEMNILAKLN ENLVELSDYV STL Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 32%.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Pseudogenes of this gene are located on more than six chromosomes including chromosome 4. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM60
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM60
Gene Accession No. Q495X7
Gene Alias 2czf45; RING finger protein 129; ring finger protein 33; RNF129; Rnf33; TRIM60; tripartite motif containing 60; tripartite motif-containing 60; tripartite motif-containing protein 60
Gene Symbols TRIM60
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM60. Recombinant protein control fragment (Product #RP-96745).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166655
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.