Learn More
Invitrogen™ TRIM60 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110522
Description
Immunogen sequence: MEFVTALVNL QEESSCPICL EYLKDPVTIN CGHNFCRSCL SVSWKDLDDT FPCPVCRFCF PYKSFRRNPQ LRNLTEIAKQ LQIRRSKRKR QKENAMCEKH NQFLTLFCVK DLEILCTQCS FSTKHQKHYI CPIKKAASYH REILEGSLEP LRNNIERVEK VIILQGSKSV ELKKKVEYKR EEINSEFEQI RLFLQNEQEM ILRQIQDEEM NILAKLNENL VELSDYVSTL KHLLREVEGK SVQSNLELLT QAKSMHHKYQ NLKCPELFSF RLTKYGFSLP PQYSGLDRII KPFQVDVILD.
The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Pseudogenes of this gene are located on more than six chromosomes including chromosome 4. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013].
Specifications
| TRIM60 | |
| Polyclonal | |
| Unconjugated | |
| TRIM60 | |
| 2czf45; RING finger protein 129; ring finger protein 33; RNF129; Rnf33; TRIM60; tripartite motif containing 60; tripartite motif-containing 60; tripartite motif-containing protein 60 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 234329 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8VI40 | |
| TRIM60 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRIM60 (NP_689833.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.