Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM61 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561884
Description
Immunogen sequence: KLEEYNAPWK ERVELIEKVI TMQTRKSLEL KKKMESPSV Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 36%.
RNF35 belongs to the TRIM/RBCC family.
Specifications
TRIM61 | |
Polyclonal | |
Unconjugated | |
TRIM61 | |
2czf61; AU018704; C85201; E330039K03Rik; putative tripartite motif-containing protein 61; RING finger protein 35; RNF35; TRIM61; tripartite motif containing 61; tripartite motif-containing 61 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
391712 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5EBN2 | |
TRIM61 | |
Recombinant protein corresponding to Human TRIM61. Recombinant protein control fragment (Product #RP-100772). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction