Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565595 Supplier Invitrogen™ Supplier No. PA565595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHEQHQVTGID DAFDELQREL KDQLQALQDS EREHTEALQL LKRQLAETKS STKSLRTTIG EAFERLHRLL RERQKA Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.

E3 ubiquitin ligase whose activity is dependent on E2 ubiquitin-conjugating enzyme UBE2D2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM62
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM62
Gene Accession No. Q9BVG3
Gene Alias 6330414G21Rik; AI450348; Dear1; ductal epithelium-associated RING Chromosome 1; E3 ubiquitin-protein ligase TRIM62; RING-type E3 ubiquitin transferase TRIM62; TRIM62; tripartite motif containing 62; tripartite motif-containing 62; tripartite motif-containing protein 62
Gene Symbols TRIM62
Host Species Rabbit
Immunogen Recombinant Human TRIM62. Recombinant protein control fragment (Product #RP-106257).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.